Learn More
STEMCELL Technologies Human Recombinant FGF-18
Click to view available options
Cantidad:
10 μg
50 μg
Tamaño de la unidad:
10 microgramos
50 microgramos
Descripción
- Fibroblast growth factor 18 (FGF-18) is a growth factor and member of the FGF subfamily
- FGF-18 is most similar to FGF-8 and FGF-17, and is secreted in adult lung and developing tissues (Ohbayashi et al.) FGF-18 signals through FGF receptor 3 (FGFR3) to regulate proliferation, differentiation, and matrix production of articular and growth plate chondrocytes in vivo and in vitro (Davidson et al.) FGF-18 has skeletal functions and protects articular cartilage by gene expression profiling and regulates proliferation and differentiation of midline cerebellar structures (Mori et al.) Also, recombinant human FGF-18 has been shown to effectively regulate hair growth (Song et al.)
Especificaciones
Specifications
| Nombre | FGFI, Fibroblast growth factor-18, zFGF5 |
| Cantidad | 10 μg |
| Tipo de célula | Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mammary Cells; Mesenchymal Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells; Prostate Cells |
| Grado de pureza o calidad | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction