Learn More
STEMCELL Technologies Human Recombinant FGF-10 (KGF-2)
Código de producto. 15892628
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1000 μg
10 μg
500 μg
50 μg
Tamaño de la unidad:
10 microgramos
1000 microgramos
50 microgramos
500 microgramos
15892628 50 μg 50 microgramos
17139411 10 μg 10 microgramos
17149411 500 μg 500 microgramos
17159411 1000 μg 1000 microgramos
4 options
Código de producto. 15892628
Marca: STEMCELL Technologies 78037.1
Fibroblast growth factor 10
  • Fibroblast growth factor 10 (FGF-10) is a member of the fibroblast growth factor (FGF) family predominantly expressed by mesenchymal fibroblasts during embryonic development (Emoto et al.; Igarashi et al.) It binds with high affinity to fibroblast growth factor receptor 2-IIIb (FGFR2-IIIb), and also has a weaker affinity for FGFR1-IIIb (Beer et al.) FGF-10 and FGF-7 have similar receptor binding properties and target cell specificities but are differentially regulated by components of the extracellular matrix (Emoto et al.; Igarashi et al.) FGF-10 has been shown to mediate epithelial-mesenchymal interactions, which are essential to lung development (Sekine et al; Ware & Matthay)
  • FGF-10 also has a role in mobilization and proliferation of lung-resident mesenchymal stem cells (MSCs) and protection and repair against acute lung injury (Tong et al.; Ware & Matthay), as well as endodermal differentiation of human pluripotent stem cells to insulin-producing pancreatic-like cells (Takeuchi et al.)
Especificaciones
Nombre FGFA, Fibroblast growth factor-10, Keratinocyte growth factor-2
Cantidad 50 μg
Tipo de célula Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Keratinocytes; Mammary Cells; Mesenchymal Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells; Prostate Cells
Grado de pureza o calidad ≥95%