Learn More
STEMCELL Technologies Human Recombinant Betacellulin
Código de producto. 17129581
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1000 μg
10 μg
50 μg
Tamaño de la unidad:
10 microgramos
1000 microgramos
50 microgramos
17129581 1000 μg 1000 microgramos
15763737 50 μg 50 microgramos
17119581 10 μg 10 microgramos
3 options
Código de producto. 17129581
Marca: STEMCELL Technologies 78105.2
Betacellulin
  • Betacellulin is a member of the epidermal growth factor (EGF) family, and signals through EGF receptor and ERBB4
  • It activates ERK and AKT pathways, which induces neural stem cell proliferation and prevents spontaneous differentiation in culture
  • Betacellulin stimulates the expansion of neural stem cells, transit-amplifying cells, and neuroblasts derived from subventricular zone and dentate gyrus
  • It is a potent mitogen for retinal pigment epithelial cells and vascular smooth muscle cells
  • Betacellulin down-regulates E-cadherin expression in ovarian cancer cell lines via MEK/ERK1/2 and PI3K/AKT signaling pathways, thus increasing cell migration
  • It is a modulator of interferon (IFN) response and enhances anti-viral effects of IFN
  • Betacellulin is expressed in pancreatic α cells, β cells, and duct cells
Especificaciones
Cantidad 1000 μg
Tipo de célula Endoderm, PSC-Derived; Neural Stem and Progenitor Cells
Grado de pureza o calidad ≥95%