Learn More
STEMCELL Technologies Human Recombinant BDNF, ACF
Código de producto. 17149641
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1000 μg
100 μg
10 μg
Tamaño de la unidad:
10 microgramos
100 microgramos
1000 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
17149641 10 μg 10 microgramos
17159641 100 μg 100 microgramos
17169641 1000 μg 1000 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 17149641 Proveedor STEMCELL Technologies N.º de proveedor 78133
Brain-derived neurotrophic factor, animal component-free
  • Brain-derived neurotrophic factor (BDNF), like nerve growth factor (NGF), neurotrophin-3 (NT-3), and neurotrophin-4 (NT-4), is a member of the NGF family of neurotrophins, which are required for the differentiation and survival of specific neuronal subpopulations in both the central and the peripheral nervous systems
  • BDNF binds with high affinity to the TRKB kinase receptor, and activates AKT and ERK pathways
  • It is expressed in hippocampus, cortex, and synapses of the basal forebrain
  • BDNF acts as a survival factor for human embryonic stem cells when plated on either feeder cells or Corning™ Matrigel™
Especificaciones
Nombre Abrineurin, ANON2, BULN2, Neurotrophin, MGC34632
Cantidad 10 μg
Tipo de célula Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons
Grado de pureza o calidad ≥95%