missing translation for 'onlineSavingsMsg'
Learn More

Human CCL5 (RANTES), Recombinant Protein, Invitrogen™

Código de producto. 11898811
Change view
Click to view available options
Cantidad:
20 μg
Tamaño de la unidad:
20 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11898811 20 μg 20 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11898811 Proveedor Invitrogen N.º de proveedor RRNTS20

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Human RANTES Recombinant Protein, RRNTS20, from Invitrogen.

This gene is one of several CC cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene functions as a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. This cytokine is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor CCR5 and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor. RRNTS20 contains 68 amino acid residues. Purity: >98% pure by SDS-PAGE analyses. Activity: Determined by its ability to chemoattract human blood monocytes using a concentration range of 1.0-10ng/mL. Reconstitution and Dilution: Reconstitute in distilled water to a concentration of 100 ug/ml. After reconstitution, dilute cytokine in a neutral buffer containing carrier protein such as 5-10% FBS or 1-4% high purity BSA (fatty acid free fraction V). Lyophilized product is stable until expiration if stored desiccated below 0°C. Cytokines diluted as indicated above should be divided into aliquots and stored at -70°C. Recombinant proteins should be stored at concentrations >500ng/ml to reduce loss of activity.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P13501
Para utilizar con (aplicación) Control, ELISA, Immunoprecipitation, Western Blot
Formulación protein with no preservative
ID de gen (Entrez) 6352
Nombre Human CCL5 (RANTES)
Cantidad 20 μg
Fuente E. Coli
Requisitos de almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Alias de gen CCL5, C-C motif chemokine 5; SIS-delta; T-cell specific protein p288; T-cell-specific protein RANTES; beta-chemokine RANTES; eoCP; eosinophil chemotactic cytokine; regulated upon activation; normally T-expressed; and presumably secreted; small inducible cytokine A5 (RANTES); small inducible cytokine subfamily A (Cys-Cys); member 5; small-inducible cytokine A5; t cell-specific protein P228
Nombre común Human CCL5 (RANTES)
Símbolo de gen CCL5
Reactividad específica Human
Conjugado Unconjugated
Reactividad cruzada Hu
Especie E. coli
Recombinante Recombinant
Formulario Lyophilized
Subtipo de proteína Recombinant
Grado de pureza o calidad purified
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.