missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Pleiotrophin (PTN) Recombinant Protein

Código de producto. 10487253
Change view
Click to view available options
Cantidad:
20 μg
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
10487253 20 μg cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10487253 Proveedor Gibco™ N.º de proveedor PHC7075

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute Pleiotrophin in sterile, distilled water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: The lyophilized protein is stable at room temperature for 1 month, but should be kept at -20°C for long term storage. Working aliquots stored with a carrier protein are stable for at least 3 months at -20°C to -80°C. Avoid repeated freeze/thaw cycles.

Pleiotrophin (PTN) is an 18 kDa secreted heparin and chondroitin-binding protein that is a regulatory cytokine critical to the early development of blood vessels and neurons. High PTN serum levels are associated with a variety of solid tumors. It was shown that patients with multiple myeloma (MM) also have elevated serum levels of this protein. The expression of this protein is very limited in normal adult tissues.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P21246
Para utilizar con (aplicación) Bioactivity
Formulación 5mM sodium phosphate with no preservative; pH 7.4
ID de gen (Entrez) 5764
Peso molecular 15.4 kDa
Nombre Human Pleiotrophin (PTN)
Intervalo de pH 7.4
Método de purificación Purified
Cantidad 20 μg
Requisitos de almacenamiento -80°C
Estado normativo RUO
Concentración de endotoxinas <0.1 ng/μg
Alias de gen HARP; HBBM; HBBN; HB-GAM; HBGF8; HBGF-8; HBNF; HBNF1; HBNF-1; heparin affin regulatory peptide; heparin affin regulatory protein; heparin binding factor; heparin-binding brain mitogen; Heparin-binding growth factor 8; heparin-binding growth-associated molecule; heparin-binding neurite outgrowth promoting factor; Heparin-binding neurite outgrowth-promoting factor; Heparin-binding neurite outgrowth-promoting factor 1; heparin-binding neurite promoting factor; heparin-binding neurotorphic factor; heparin-binding neutrophic factor; NEGF1; OSF; Osf1; OSF-1; osteoblastic cell factor; Osteoblast-specific factor 1; pleiotrophin; pleiotrophin (heparin binding growth factor 8, neurite growth-promoting factor 1); Pleiotrophin (Heparine binding factor, Hbnf, in the mouse); PTN
Nombre común Pleiotrophin
Símbolo de gen PTN
Tipo de producto Protein
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Secuencia Human Pleiotrophin recombinant protein contains 136 amino acids
Contenido y almacenamiento -80°C
Nivel de endotoxinas <0.1 ng/μg
Condiciones de envío Approved for shipment on Wet or Dry Ice
Sistema de expresión E. coli
Familia de proteínas Cytokines & Receptors
Forma de proteína Recombinant, Ligand
Formulario Lyophilized
Subtipo de proteína Other Proteins
Categoría de investigación Stem Cell Research, Neurobiology
Línea de productos Gibco
Grado de pureza o calidad >98% by SDS-PAGE
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.