missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PDC (aa 28-80) Control Fragment Recombinant Protein
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66472 (PA5-66472. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
PDC encodes a phosphoprotein, which is located in the outer and inner segments of the rod cells in the retina. This protein may participate in the regulation of visual phototransduction or in the integration of photoreceptor metabolism. It modulates the phototransduction cascade by interacting with the beta and gamma subunits of the retinal G-protein transducin. PDC is a potential candidate gene for retinitis pigmentosa and Usher syndrome type II. Alternatively spliced transcript variants encoding different isoforms have been identified.
Especificaciones
Especificaciones
| Número de acceso | P20941 |
| Concentración | ≥5.0 mg/mL |
| Para utilizar con (aplicación) | Blocking Assay, Control |
| Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
| ID de gen (Entrez) | 5132 |
| Nombre | Human PDC (aa 28-80) Control Fragment |
| Cantidad | 100 μL |
| Estado normativo | RUO |
| Alias de gen | 33 kDa phototransducing protein; 33 DPTP; G beta gamma binding protein; MEKA; MEKA protein; Pdc; PHD; PhLOP; PhLP; Phosducin; phosducin-like orphan protein; Phototransducing protein, 33 kDa (phosducin); Protein MEKA; Rod photoreceptor 1; Rpr1; RPR-1 |
| Nombre común | PDC |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido