missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PCDHA8 (aa 774-814) Control Fragment Recombinant Protein
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (34%), Rat (34%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60763 (PA5-60763. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined.
Especificaciones
Especificaciones
| Número de acceso | Q9Y5H6 |
| Concentración | ≥5.0 mg/mL |
| Para utilizar con (aplicación) | Blocking Assay, Control |
| Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
| ID de gen (Entrez) | 56140 |
| Nombre | Human PCDHA8 (aa 774-814) Control Fragment |
| Cantidad | 100 μL |
| Estado normativo | RUO |
| Alias de gen | KIAA0345-like 6; PCDHA8; PCDH-ALPHA8; PCDH-alpha-8; protocadherin alpha 8; protocadherin alpha-8 |
| Nombre común | PCDHA8 |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido