missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human P2Y13 (aa 23-47) Control Fragment Recombinant Protein

Código de producto. 30194370
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30194370 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30194370 Proveedor Invitrogen™ N.º de proveedor RP95328

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111286 (PA5-111286. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is activated by ADP.
TRUSTED_SUSTAINABILITY

Specifica

Número de acceso Q9BPV8
Concentración ≥5.0 mg/mL
Para utilizar con (aplicación) Blocking Assay, Control
Formulación 1 M urea, PBS with no preservative; pH 7.4
ID de gen (Entrez) 53829
Nombre Human P2Y13 (aa 23-47) Control Fragment
Cantidad 100 μL
Estado normativo RUO
Alias de gen 2010001L06Rik; FKSG77; G protein-coupled receptor 68; G protein-coupled receptor 86; GPCR1; GPR86; GPR94; G-protein coupled receptor 86; G-protein coupled receptor 94; P2ry13; P2Y purinoceptor 13; P2Y13; P2yr13; purinergic receptor P2Y G-protein coupled 13; purinergic receptor P2Y, G-protein coupled 13; purinergic receptor P2Y, G-protein coupled, 13; purinergic receptor P2Y13; SP174; Vpurinergic receptor P2Y, G-protein coupled 13
Nombre común P2Y13
Símbolo de gen P2RY13
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína His-ABP-tag
Secuencia NTTVMQGFNRSERCPRDTRIVQLVF
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Sistema de expresión E. coli
Formulario Liquid
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.