missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human p16-INK4a Recombinant Protein, PeproTech®

Código de producto. 17810123
Change view
Click to view available options
Cantidad:
10 x 100 μg
100 μg
20 μg
5 x 100 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
5 microgramos
500 microgramos
paquete de 10
paquete de 5
Código de producto. Cantidad unitSize
17810123 5 x 100 μg 500 microgramos
17811663 100 μg 100 microgramos
17800573 10 x 100 μg 1 miligramo
30088488 10 x 100 μg paquete de 10
17890693 20 μg 20 microgramos
30088482 5 x 100 μg paquete de 5
17830023 5 μg 5 microgramos
7 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17810123

Marca: Gibco™ 11002500UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

110-02-1MG will be provided as 10 x 100 μg (110-02-100UG). 110-02-500UG will be provided as 5 x 100 μg (110-02-100UG). Recombinant Human p16-INK4a is a 16.5 kDa protein containing 156 amino acid residues. Kim, W.Y. and Sharpless, N.E. (2006)Cell 127(2):265-75. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene generates several transcript variants which differ in their first exons. At least three alternatively spliced variants encoding distinct proteins have been reported, two of which encode structurally related isoforms known to function as inhibitors of CDK4 kinase. The remaining transcript includes an alternate first exon located 20 Kb upstream of the remainder of the gene; this transcript contains an alternate open reading frame (ARF) that specifies a protein which is structurally unrelated to the products of the other variants. This ARF product functions as a stabilizer of the tumor suppressor protein p53 as it can interact with, and sequester, the E3 ubiquitin-protein ligase MDM2, a protein responsible for the degradation of p53. In spite of the structural and functional differences, the CDK inhibitor isoforms and the ARF product encoded by this gene, through the regulatory roles of CDK4 and p53 in cell cycle G1 progression, share a common functionality in cell cycle G1 control. This gene is frequently mutated or deleted in a wide variety of tumors, and is known to be an important tumor suppressor gene.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P42771
Para utilizar con (aplicación) Functional Assay
Formulación protein with no preservative
ID de gen (Entrez) 1029
Peso molecular 16.5
Nombre Human p16-INK4a
Cantidad 5 x 100 μg
Fuente E. coli
Requisitos de almacenamiento -20°C
Estado normativo RUO
Concentración de endotoxinas <1 EU/μg
Alias de gen Alternative reading frame; Arf; ARF-INK4a; CDK4 inhibitor p16 INK4; CDK4 inhibitor p16-INK4; CDK4I; CDKN2; Cdkn2a; cdkn2a {ECO:0000312; cell cycle inhibitor; cell cycle negative regulator beta; cell cycle regulator; CMM2; Cyclin dependent kinase 4 inhibitor A; cyclin dependent kinase inhibitor 2 A; Cyclin dependent kinase inhibitor 2 A (p16, inhibits CDK4); cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2 A; cyclin-dependent kinase inhibitor 2 A (melanoma, p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2 A (p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2 A p16Ink4a; cyclin-dependent kinase inhibitor 2 A p19Arf; cyclin-dependent kinase inhibitor 2 A, isoform 1; cyclin-dependent kinase inhibitor 2 A, isoform 2; cyclin-dependent kinase inhibitor 2 A, isoform 3; cyclin-dependent kinase inhibitor 2 A, isoforms 1/2; cyclin-dependent kinase inhibitor protein; EMBL:AAL76336.1, ECO:0000312; EMBL:AAL76338.1, ECO:0000312; HGNC:1787; inhibits CDK4; INK4; INK4A; Ink4a/Arf; INK4a-ARF; isoforms 1/2/3; Melanoma p16 inhibits CDK4; mitochondrial smARF; MLM; MTS1; MTS-1; Multiple tumor suppressor 1; P14; P14 P16; P14ARF; p16; p16(INK4a); p16Cdkn2a; P16INK4; p16-INK4; p16INK4a; p16-INK4a; P19; P19 TP16; p19; P19ARF; Pctr1; RGD:2323}; TP16; Tumor suppressor ARF
Símbolo de gen CDKN2A
Reactividad específica Human
Conjugado Unconjugated
Recombinante Recombinant
Secuencia MEPAAGSSME PSADWLATAA ARGRVEEVRA LLEAGALPNA PNSYGRRPIQ VMMMGSARVA ELLLLHGAEP NCADPATLTR PVHDAAREGF LDTLVVLHRA GARLDVRDAW GRLPVDLAEE LGHRDVARYL RAAAGGTRGS NHARIDAAEG PSDIPD
Formulario Lyophilized
Grado de pureza o calidad ≥ 95% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.