missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NGF Recombinant Protein

Código de producto. 17144243
Change view
Click to view available options
Cantidad:
5 x 50 μg
5 x 5 μg
50 μg
5 μg
Tamaño de la unidad:
5 microgramos
50 microgramos
paquete de 5
Código de producto. Cantidad unitSize
17144243 5 x 50 μg paquete de 5
17154243 5 μg 5 microgramos
17134243 5 x 5 μg paquete de 5
17164243 50 μg 50 microgramos
4 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17144243

Marca: Invitrogen™ A42576

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Nerve growth factor (NGF) influences the survival and differentiation of a variety of neuronal and nonneuronal cells. The actions of this polypeptide result from binding to specific cell surface receptors which are present as both high and low affinity sites. Although both types of receptor bind NGF only the high affinity form is capable of signal transduction. Cross-linking studies with 125I-NGF have shown that the high affinity receptor of rat PC12 cells forms an Mr 158,000 complex with NGF while the low affinity receptor forms an Mr 100,000 complex. Full length cDNA clones for both the human and rat NGF receptor genes have been produced and sequenced, revealing greater than 90% homology between the two species. NGF exhibits nerve growth stimulating activity and is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons. Further, NGF stimulates division and differentiation of sympathetic and embryonic sensory neurons. Mutations in the NGF gene have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of NGF gene expression is associated with allergic rhinitis.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P01138
Para utilizar con (aplicación) Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot
Formulación 50 mM sodium acetate with 0.02% Tween 80, 150 mM NaCl, 5% mannitol, 5% trehalose and no preservative; pH 5.5
ID de gen (Entrez) 4803
Nombre Human NGF
Cantidad 5 x 50 μg
Fuente CHO cells
Requisitos de almacenamiento -20°C
Estado normativo RUO
Concentración de endotoxinas <1 EU/μg
Alias de gen B nerve growth factor; beta ngf; beta-nerve growth factor; BetaNGF; beta-NGF; bNGF; b-NGF; HSAN5; MGC161426; MGC161428; nerve growth factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta subunit; NGF; NGF b; NGF β; NGFB; NGFbeta; NGFβ; β nerve growth factor; β NGF; βNGF
Símbolo de gen NGF
Reactividad específica Human
Conjugado Unconjugated
Recombinante Recombinant
Formulario Lyophilized
Grado de pureza o calidad ≥ 95%
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.