missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NGF Recombinant Protein

Código de producto. 17935731
Click to view available options
Cantidad:
100 μg
1 mg
Tamaño de la unidad:
1 miligramo
100 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17935731

Marca: Invitrogen™ RP88189

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. Reconstituted with sterile deionized water to 0.59mg/mL. Concentration is measured by UV.

Nerve growth factor (NGF) influences the survival and differentiation of a variety of neuronal and nonneuronal cells. The actions of this polypeptide result from binding to specific cell surface receptors which are present as both high and low affinity sites. Although both types of receptor bind NGF only the high affinity form is capable of signal transduction. Cross-linking studies with 125I-NGF have shown that the high affinity receptor of rat PC12 cells forms an Mr 158,000 complex with NGF while the low affinity receptor forms an Mr 100,000 complex. Full length cDNA clones for both the human and rat NGF receptor genes have been produced and sequenced, revealing greater than 90% homology between the two species. NGF exhibits nerve growth stimulating activity and is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons. Further, NGF stimulates division and differentiation of sympathetic and embryonic sensory neurons. Mutations in the NGF gene have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of NGF gene expression is associated with allergic rhinitis.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P01138
Concentración 0.59 mg/mL
Para utilizar con (aplicación) Control, Bioactivity
Formato Lyophilized
Formulación 40mM arginine, 40mM histidine with 5% mannitol, 150mM NaCl, 5% trehalose, 0.01% Tween 80 and no preservative; pH 5.5
ID de gen (Entrez) 4803
Peso molecular 13.27 kDa
Nombre Human NGF
Intervalo de pH 5.5
Método de purificación Purified
Cantidad 100 μg
Requisitos de almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Concentración de endotoxinas <1.0 EU/μg
Alias de gen B nerve growth factor; beta ngf; beta-nerve growth factor; BetaNGF; beta-NGF; bNGF; b-NGF; HSAN5; MGC161426; MGC161428; Nerve growth factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta polypeptide; nerve growth factor, beta subunit; NGF; NGF b; NGF β; Ngfb; NGFbeta; NGFβ; β nerve growth factor; β NGF; βNGF
Nombre común NGF
Símbolo de gen NGF
Actividad biológica Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. The ED50 for this effect is typically 0.2-2 ng/mL
Tipo de producto Protein
Conjugado Unconjugated
Recombinante Recombinant
Secuencia Mature form of human beta-NGF, amino acids Ser122-Arg239 (Accession # NP_002497.2)
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Actividad Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. The ED50 for this effect is typically 0.2-2 ng/mL.
Sistema de expresión CHO cells
Formulario Lyophilized
Grado de pureza o calidad ≥95% as determined by SDS-PAGE.
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado