missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neutrophil Elastase/ELA2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB91673-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Especificaciones
| Neutrophil Elastase/ELA2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2, ELA2granulocyte-derived elastase, ELANE, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, Leukocyte Elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Elastase/ELA-2, Met1-Asn252, Accession # P08246 | |
| 100 μg | |
| 1991 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA, Flow Cytometry, CyTOF | |
| 950312 | |
| ELISA, Intracellular Staining by Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| P08246 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Elastase/ELA-2 in direct ELISAs. | |
| Human | |
| Lyophilized |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido