missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human MIP-1 beta (CCL4) Recombinant Protein, PeproTech®

Código de producto. p-7246204
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
250 μg
2 μg
500 μg
50 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
2 microgramos
250 microgramos
50 microgramos
500 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17801703

Marca: Gibco™ 30009100UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Human MIP-1beta is a 7.6 kDa protein containing 69 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CCL4 (macrophage inflammatory protein 1-beta, MIP1B) belongs to the intercrine beta (chemokine CC) family. Functionally, CCL4 is involved in chemotactic and proinflammatory effects, and homeostasis. CCL4 is produced by macrophages upon stimulation by bacterial endotoxins. CCL4 recruits and stimulates various inflammatory cells at sites of inflammation. CCL4 is produced by lymphocytes, macrophages and dendritic cells. Both CCL4 and the related protein CCL3 participate in the host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells. While both CCL4 and CCL3 exert similar effects on monocytes, their effect on lymphocytes differ, with CCL4 selectively attracting CD4+ lymphocytes and CCL3 selectively attracting CD8+ lymphocytes. Additionally, both have been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. The processed form of CCL4 can induce down-modulation of surface expression of the chemokine receptor CCR5, thus inhibiting the CCR5-mediated entry of HIV-1 in T cells. CCL4 binds with high affinity to CCR5 receptors.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P13236
Para utilizar con (aplicación) ELISA, Functional Assay, Western Blot
Formulación protein with no preservative
ID de gen (Entrez) 6351
Peso molecular 7.6 kDa
Nombre Human MIP-1 beta (CCL4)
Cantidad 100 μg
Fuente E. coli
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen ACT2; ACT-2; AT744.1; CC chemokine ligand 4; C-C motif chemokine 4; C-C motif chemokine ligand 4; Ccl4; chemokine (C-C motif) ligand 4; G-26; G-26 T-lymphocyte-secreted protein; HC21; immune activation gene 2; immune activation gene-2; immune activation protein 2; LAG1; LAG-1; Lymphocyte activation gene 1 protein; macrophage inflammatory protein 1-beta; macrophage inflammatory protein-1 beta; MGC104418; MGC126025; MGC126026; MIP-1 beta; MIP1b; MIP-1 B; Mip1-b; MIP1B1; MIP1beta; MIP-1 BETA; MIP-1-beta; MIP1-beta; MIP-1-beta(1-69); MIP-1-beta(3-69); PAT 744; Protein H400; putative MIP-1 beta protein; RP23-320E6.8; SCY4A; SCYA2; Scya4; secreted protein G-26; SIS-gamma; small inducible cytokine A4; small inducible cytokine A4 (homologous to mouse Mip-1 b); small-inducible cytokine A4; T-cell activation protein 2
Nombre común CCL4
Símbolo de gen CCL4
Actividad biológica Determined by its ability to chemoattract human blood monocytes using a concentration range of 5.0-20.0 ng/ml
Conjugado Unconjugated
Recombinante Recombinant
Secuencia APMGSDPPTA CCFSYTARKL PHNFVVDYYE TSSLCSQPAV VFQTKRGKQV CADPSESWVQ EYVYDLELN
Contenido y almacenamiento -20°C
Sistema de expresión E. coli
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado