missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Midkine (MK) Recombinant Protein

Código de producto. 10532825
Change view
Click to view available options
Cantidad:
20 μg
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
10532825 20 μg cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10532825 Proveedor Gibco™ N.º de proveedor PHC7084

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. This lyophilized protein should be reconstituted in sterile, distilled water to a concentration of 0.1-1 mg/mL. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C.

Midkine is a retinoic acid-responsive, heparin-binding growth factor expressed in various cell types during embryogenesis. It promotes angiogenesis, cell growth, and cell migration. Midkine is also expressed in several carcinomas, suggesting that it may play a role in tumorigenesis, perhaps through its effects on angiogenesis.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P21741
Para utilizar con (aplicación) Bioactivity
Formulación PBS with no preservative
ID de gen (Entrez) 4192
Peso molecular 13.4 kDa
Nombre Human Midkine (MK)
Método de purificación Purified
Cantidad 20 μg
Requisitos de almacenamiento -20°C
Estado normativo RUO
Concentración de endotoxinas <0.1 ng/μg
Alias de gen amphiregulin-associated protein; ARAP; FLJ27379; MDK; Mek; Midgestation and kidney protein; midkine; midkine (neurite growth-promoting factor 2); MK; MK1; NEGF2; Neurite outgrowth-promoting factor 2; Neurite outgrowth-promoting protein; Retanoic acid-responsive protein; retinoic acid inducible factor; retinoic acid-induced differentiation factor; retinoic acid-responsive protein
Nombre común Midkine
Símbolo de gen Mdk
Actividad biológica ED50 = 0.1 - 10 ng/mL; determined by the dose-dependent chemotaxis of human neutrophils.
Tipo de producto Protein
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Secuencia Human Midkine recombinant protein contains 123 amino acids
Contenido y almacenamiento -20°C
Clasificación Carrier-Free
Actividad ED50 = 0.1 - 10 ng/mL; determined by the dose-dependent chemotaxis of human neutrophils.
Nivel de endotoxinas <0.1 ng/μg
Condiciones de envío Approved for shipment on Wet or Dry Ice
Sistema de expresión E. coli
Familia de proteínas Chemokines & Receptors
Forma de proteína Recombinant, Ligand
Formulario Lyophilized
Subtipo de proteína Other Proteins
Categoría de investigación Neurobiology, Oncology, Stem Cell Research, Cardiovascular Research, Differentiation, Angiogenesis
Línea de productos Gibco
Grado de pureza o calidad >98% by SDS-PAGE
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.