missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human MEC (CCL28) Recombinant Protein, PeproTech®

Product Code. 17812553
Change view
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
250 μg
500 μg
5 μg
Unit Size:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
5 microgramos
500 microgramos
Product Code. Cantidad unitSize
17812553 500 μg 500 microgramos
17832653 100 μg 100 microgramos
17862553 1 mg 1 miligramo
17802763 20 μg 20 microgramos
17852263 250 μg 250 microgramos
17852653 5 μg 5 microgramos
6 options
This item is not returnable. View return policy

Product Code. 17812553

Brand: Gibco™ 30057500UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human MEC is a 12.3 kDa protein containing 108 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

MEC is a secreted CC chemokine expressed primarily by epithelial cells of the bronchioles, salivary gland, mammary gland and colon. MEC signals through the CCR10 receptor, and chemoattracts resting CD4, CD8 T-cells and eosinophils. MEC contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines.
TRUSTED_SUSTAINABILITY

Specifications

Número de acceso Q9NRJ3
Para utilizar con (aplicación) ELISA, Functional Assay, Western Blot
Formulación protein with no preservative
ID de gen (Entrez) 56477
Peso molecular 12.3 kDa
Nombre Human MEC (CCL28)
Cantidad 500 μg
Fuente E. coli
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen CC chemokine CCL28; C-C motif chemokine 28; C-C motif chemokine ligand 28; CCK 1; CCK1; CCK1 protein; CCL 28; Ccl28; chemokine (C-C motif) ligand 28; chemokine (C-C motif) ligand 28 splice variant chi; MEC; Mucosae-associated epithelial chemokine; Protein CCK1; SCYA28; small inducible cytokine A28; small inducible cytokine subfamily A (Cys-Cys), member 28; small-inducible cytokine A28
Nombre común CCL28
Símbolo de gen CCL28
Actividad biológica Determined by its ability to chemoattract human lymphocytes using a concentration range of 1.0-10.0 ng/ml
Conjugado Unconjugated
Recombinante Recombinant
Secuencia SEAILPIASS CCTEVSHHIS RRLLERVNMC RIQRADGDCD LAAVILHVKR RRICVSPHNH TVKQWMKVQA AKKNGKGNVC HRKKHHGKRN SNRAHQGKHE TYGHKTPY
Contenido y almacenamiento -20°C
Sistema de expresión E. coli
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.