missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LAIR2 Recombinant Protein
Descripción
It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (599 μL) be added to the vial to prepare a stock solution of 1.67 mg/mL. Concentration is measured by UV.
The protein encoded by this gene is a member of the immunoglobulin superfamily. It was identified by its similarity to leukocyte-associated immunoglobulin-like receptor 1, a membrane-bound receptor that modulates innate immune response. The protein encoded by this locus is a soluble receptor that may play roles in both inhibition of collagen-induced platelet aggregation and vessel formation during placental implantation. This gene maps to a region of 19q13.4, termed the leukocyte receptor cluster, which contains 29 genes in the immunoglobulin superfamily. Alternatively spliced transcript variants have been described for this gene.
Especificaciones
Especificaciones
| Número de acceso | Q6ISS4 |
| Concentración | 0.25 mg/mL |
| Para utilizar con (aplicación) | Control, Neutralization |
| Formato | Lyophilized |
| Formulación | 50mM tris with 5% mannitol, 0.7mM NaCl, 5% trehalose and no preservative; pH 8 |
| ID de gen (Entrez) | 3904 |
| Peso molecular | 22 kDa |
| Nombre | Human LAIR2 |
| Intervalo de pH | 8 |
| Método de purificación | Purified |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?