missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LAIR1 Fc Chimera Recombinant Protein

Código de producto. 17906151
Change view
Click to view available options
Cantidad:
1 mg
50 μg
Tamaño de la unidad:
1 miligramo
50 microgramos
Código de producto. Cantidad unitSize
17906151 50 μg 50 microgramos
17916151 1 mg 1 miligramo
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17906151

Marca: Invitrogen™ RP88099

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (1000 μL) be added to the vial to prepare a stock solution of 1 mg/mL. Concentration is measured by UV.

CD305, also known as LAIR1, is a type I transmembrane glycoprotein with an extracellular Ig-like domain and two intracellular ITIMs. It is expressed on a wide range of immune cells, including T, NK, and B cells, monocytes, dendritic cells, eosinophils, basophils, and mast cells. In humans, CD305 is also present on CD34+ hematopoietic progenitor cells and thymocytes. Several isoforms exist due to alternative splicing, with expression varying between cell types. CD305 functions as an inhibitory receptor, playing a key role in regulating immune responses. It blocks cytokine-mediated signaling, cell proliferation, and apoptosis. In NK, B, and T cells, CD305 modulates cytolytic functions and cytokine production, down-regulating IL2 and IFNG in CD4+ T cells while inducing transforming growth factor beta. In B cells, it reduces IgG and IgE production and decreases IL8, IL10, and TNF secretion. Activation of CD305 through tyrosine phosphorylation recruits phosphatases PTPN6 and PTPN11, although it may also function independently of SH2-containing phosphatases. It inhibits differentiation of peripheral blood precursors into dendritic cells and has been shown to induce apoptosis in myeloid leukemia cell lines. Collagens are identified as ligands for CD305, underscoring its role in immune regulation.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso Q6GTX8
Concentración 0.25 mg/mL
Para utilizar con (aplicación) Control, Neutralization
Formato Lyophilized
Formulación PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
ID de gen (Entrez) 3903
Peso molecular 42.8 kDa
Nombre Human LAIR1 Fc Chimera
Intervalo de pH 7.4
Método de purificación Purified
Cantidad 50 μg
Requisitos de almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Concentración de endotoxinas <1.0 EU/μg
Alias de gen 5133400O11Rik; BB115266; CD305; D7Bwg0421e; immunoglobulin heavy chain variable region; Lair1; Lair-1; leukocyte associated immunoglobulin like receptor 1; leukocyte-associated Ig-like receptor 1; leukocyte-associated Ig-like receptor-1; leukocyte-associated immunoglobulin-like receptor 1; rLAIR1
Nombre común LAIR1 (CD305)
Símbolo de gen LAIR1
Tipo de producto Protein
Conjugado Unconjugated
Recombinante Recombinant
Etiqueta de proteína Fc-tag
Secuencia Human LAIR1, amino acids Met1-Tyr165 (Accession # Q6GTX8.1) with a C-terminal human IgG1 Fc region
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Sistema de expresión HEK293 cells
Formulario Lyophilized
Grado de pureza o calidad >95% as determined by SDS-PAGE.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.