missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Marca: R&D Systems MAB207-500
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IL-7 Monoclonal specifically detects IL-7 in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Especificaciones
| IL-7 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 0.4-0.8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P13232 | |
| IL7 | |
| E. coli-derived recombinant human IL-7 Accession # P13232 | |
| 500 μg | |
| Primary | |
| Detects human IL-7 in Western blots. In ELISAs, does not cross-react with bovine FGF acidic, recombinant human (rh) G-CSF, rhGM-CSF, rhIL-1 alpha , rhIL-1 beta , rhIL-2, rhIL-3, rhIL-4, rhIL-6, recombinant mouse IL-7, rhIL-8, porcine PDGF, rhTNF-alpha , rhTNF-beta , human TGF-beta 1, or porcine TGF-beta 1. | |
| Human | |
| Ascites |
| Western Blot, Neutralization, ELISA | |
| 7417 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL7, IL-7interleukin-7, interleukin 7, Lymphopoietin-1, PBGF | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3574 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido