missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-6R (soluble) Recombinant Protein, PeproTech®

Código de producto. 17821673
Change view
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
250 μg
500 μg
50 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
5 microgramos
50 microgramos
500 microgramos
Código de producto. Cantidad unitSize
17821673 500 μg 500 microgramos
17891183 100 μg 100 microgramos
17890503 1 mg 1 miligramo
17830713 20 μg 20 microgramos
17861193 250 μg 250 microgramos
17851843 50 μg 50 microgramos
17801073 5 μg 5 microgramos
7 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17821673

Marca: Gibco™ 20006RC500UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human sIL-6 Receptor alpha is a 37.9 kDa glycoprotein corresponding to 339 amino acid residues of the extracellular domain of IL-6Ralpha. As a result of glycosylation, Recombinant Human sIL-6 Receptor alpha migrates with an apparent molecular mass of approximately 57-70 kDa by SDS-PAGE gel, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 6 receptor (IL6R), also known as CD126 (Cluster of Differentiation 126), is a type I cytokine receptor. This gene encodes a subunit of the interleukin 6 (IL6) receptor complex. Interleukin 6 is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in the immune response. The IL6 receptor is a protein complex consisting of this protein and interleukin 6 signal transducer (IL6ST/GP130/IL6-beta), a receptor subunit also shared by many other cytokines. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer. Alternatively spliced transcript variants encoding distinct isoforms have been reported. A pseudogene of this gene is found on chromosome 9.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P08887
Para utilizar con (aplicación) Functional Assay
Formulación protein with no preservative
ID de gen (Entrez) 3570
Peso molecular 37.9 kDa
Nombre Human IL-6 R (soluble)
Cantidad 500 μg
Fuente CHO cells
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen CD126; CD126 antigen; gp80; IL-6 Receptor; il-6 receptor alpha; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL6Q; IL6R; IL-6 R; IL-6 R 1; IL-6 R subunit alpha; IL6R1; IL-6 R-1; Il6ra; IL-6 RA; IL-6 R-alpha; IL6RQ; ILN; Interleukin; interleukin 6 receptor; interleukin 6 receptor, alpha; interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; MGC104991
Nombre común IL-6 Receptor (CD126)
Símbolo de gen IL6R
Actividad biológica Determined by its ability to intensify the IL-6 induced growth inhibition of mouse M1 cells. The expected ED50 is ≤ 5.0 ng/ml, in the presence of 20 ng/ml of rhIL-6.
Conjugado Unconjugated
Recombinante Recombinant
Secuencia LAPRRCPAQE VARGVLTSLP GDSVTLTCPG VEPEDNATVH WVLRKPAAGS HPSRWAGMGR RLLLRSVQLH DSGNYSCYRA GRPAGTVHLL VDVPPEEPQL SCFRKSPLSN VVCEWGPRST PSLTTKAVLL VRKFQNSPAE DFQEPCQYSQ ESQKFSCQLA VPEGDSSFYI VSMCVASSVG SKFSKTQTFQ GCGILQPDPP ANITVTAVAR NPRWLSVTWQ DPHSWNSSFY RLRFELRYRA ERSKTFTTWM VKDLQHHCVI HDAWSGLRHV VQLRAQEEFG QGEWSEWSPE AMGTPWTESR SPPAENEVST PMQALTTNKD DDNILFRDSA NATSLPVQD
Contenido y almacenamiento -20°C
Sistema de expresión CHO cells
Formulario Lyophilized
Grado de pureza o calidad ≥ 95% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.