missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-33 Recombinant Protein

Código de producto. 17676625
Change view
Click to view available options
Cantidad:
100 μg
10 μg
25 μg
500 μg
Tamaño de la unidad:
10 microgramos
100 microgramos
25 microgramos
500 microgramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17676625 25 μg 25 microgramos
17666625 10 μg 10 microgramos
17686625 100 μg 100 microgramos
17696625 500 μg 500 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17676625 Proveedor Invitrogen™ N.º de proveedor RP87833

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

E. coli Recombinant Protein

IL-33 (Interleukin-33) is a 270 amino acid, highly divergent protein belonging to the IL-1 family with an IL-1-like C-terminal domain. IL-33 is a dual function protein that may function both as a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. IL-33 binds to and signals through IL1RL1/ST2 and its stimulation recruits MYD88, IRAK1, IRAK4, and TRAF6. IL-33 activates NF-kappaB and MAP kinases, and drives production of TH2-associated cytokines from in vitro polarized TH2 cells. In vivo, IL-33 induces the expression of IL-4, IL-5, and IL-13 and leads to severe pathological changes in mucosal organs. IL-33 is proteolytically converted to a mature form by CASP1 and is highly expressed in high endothelial venules found in tonsils, Peyer's patches and mesenteric lymph nodes and is almost undetectable in placenta. Prolonged IL-33 treatment of mice led to the development of eosinophilia, splenomegaly, and severe pathological changes in mucosal organs such as lungs, esophagus and small intestine. Recent experiments have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. Despite its predicted molecular weight, IL-33 will often run at higher molecular weight in SDS-PAGE. Studies have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine, and an intracellular nuclear factor with transcriptional regulatory properties.
TRUSTED_SUSTAINABILITY

Spezifikation

Número de acceso O95760
Concentración 200 μg/mL
Para utilizar con (aplicación) Control, ELISA, Western Blot
Formulación PBS with 1 mM EDTA, 2 mM DTT and no preservative; pH 7.2
ID de gen (Entrez) 90865
Peso molecular 18 kD
Nombre Human IL-33 (aa 112-270)
Cantidad 25 μg
Fuente E. coli
Requisitos de almacenamiento -20°C
Estado normativo RUO
Concentración de endotoxinas 0.01 ng/ μg cytokine
Alias de gen 9230117N10Rik; C9orf26; DVS27; DVS27-related protein; H-IL-33; Il1f11; IL-1F11; Il33; IL-33; ILN; Interleukin; interleukin 33; Interleukin-1 family member 11; Interleukin33; Interleukin-33; Interleukin-33 (109-270); Interleukin-33 (95-270); Interleukin-33 (99-270); Interleukin-33(102-266); Interleukin-33(109-266); NFEHEV; NFHEV; NF-HEV; nuclear factor for high endothelial venules; nuclear factor from high endothelial venules; RGD1311155; RP11-575C20.2
Símbolo de gen IL33
Reactividad específica Human
Conjugado Unconjugated
Recombinante Recombinant
Formulario Liquid
Grado de pureza o calidad >95%
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.