missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-29 (IFN lambda-1) Recombinant Protein, PeproTech®

Código de producto. 17801003
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
250 μg
500 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
5 microgramos
500 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17801003

Marca: Gibco™ 30002L500UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Human IFN-lambda1 is a 19.8 kDa protein containing 178 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IFN-lambdas constitute a novel family of interferons that mediate the induction of anti-viral protection in a wide variety of cells. There are three members in this family, namely lambda1, lambda2, and lambda3, which are also known as IL-29, IL-28A, and IL-28B, respectively. Similar to type I IFNs, IFN-lambdas activate an intracellular signaling to promote gene expression. IFN-lambdas mediate their anti-viral protection through a class II cytokine receptor complex composed of two essential receptor proteins, CRF2-12/IFN-lambdaR1, which is unique to IFN-lambdas, and CFR2-4/IL-10R2, which is shared with IL-10, IL-22, and IL-26 receptors. The IFN-lambdas, IL-28 and IL-29, have recently been reported to prime dendritic cells to induce proliferation of Foxp3-bearing regulatory T cells. DCs that differentiate in response to IFN-lambdas express high levels of class I and II MHC gene products, but low levels of costimulatory molecules. Moreover, these cells can specifically induce IL-2-dependent proliferation of CD4+CD25+FOXP3+ T cells with contact-dependent suppressive activity.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso Q8IU54
Para utilizar con (aplicación) Functional Assay
Formulación protein with no preservative
ID de gen (Entrez) 282618
Peso molecular 19.8 kDa
Nombre Human IL-29 (IFN lambda-1)
Cantidad 500 μg
Fuente E. coli
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen cytokine Zcyto21; Cytokine Zcyto22; FNL1; IFN lambda1; IFNL1; IFNL3; IFN-lambda-1; IFN-lambda-3; IFN-lambda-4; IL28B; IL-28 B; IL28C; IL-28 C; IL29; IL-29; ILN; interferon lambda 1; interferon lambda 3; Interferon lambda1; interferon lambda-1; Interferon lambda-3; interferon, lambda 1; interferon, lambda 3; interferon, lambda 4; interferon-lambda-1; Interleukin; interleukin 29 (interferon, lambda 1); interleukin-28 B; Interleukin-28 C; interleukin-29; ZCYTO21; ZCYTO22
Nombre común IL-29
Símbolo de gen IFNL1
Actividad biológica Determined by its ability to activate STAT phosphorylation in an ISRE Luciferase Reporter Assay using human colon carcinoma COLO205 cells. The expected ED50 is 0.2-0.5 ng/ml
Conjugado Unconjugated
Recombinante Recombinant
Secuencia PTSKPTTTGK GCHIGRFKSL SPQELASFKK ARDALEESLK LKNWSCSSPV FPGNWDLRLL QVRERPVALE AELALTLKVL EAAAGPALED VLDQPLHTLH HILSQLQACI QPQPTAGPRP RGRLHHWLHR LQEAPKKESA GCLEASVTFN LFRLLTRDLK YVADGNLCLR TSTHPEST
Contenido y almacenamiento -20°C
Sistema de expresión E. coli
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado