missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-22 Recombinant Protein, PeproTech®

Código de producto. 17831703
Change view
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
250 μg
2 μg
500 μg
50 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
2 microgramos
250 microgramos
50 microgramos
500 microgramos
Código de producto. Cantidad unitSize
17831703 10 μg 10 microgramos
17880433 100 μg 100 microgramos
17830343 1 mg 1 miligramo
17830913 250 μg 250 microgramos
17800613 2 μg 2 microgramos
17800443 500 μg 500 microgramos
17871093 50 μg 50 microgramos
7 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17831703

Marca: Gibco™ 2002210UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Edge
Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Human IL-22 is a 33.6 kDa, non-disulfide-linked, homodimeric protein consisting of two 147 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-22 also known as IL-10-related T-cell derived inducible factor, is an alpha helical cytokine and is considered a member of the IFN-IL-10 family, which includes IL-19, IL-20, IL-24, IL-26, IL-28, IL-29, and the type I and II interferons. IL-22 is produced mainly by activated T cells and NK cells. In humans, the IL-22 gene is located on the q arm of chromosome 12, and is structurally related to IL10. IL-22 acts by engaging the heterodimeric receptor complex consisting of primary receptor IL-22R1 and accessory receptor IL-10R2. IL-22R1 also binds IL-20 and IL-24; IL-10R2 also binds IL-10, IL-27, IL-28, and IL-29. Binding of IL-22 to its receptor complex induces signal transduction, particularly via the JAK-STAT pathway. In addition to the membrane-bound IL-22R1/IL-10R2 complex, a soluble single-chain IL-22 receptor termed IL-22BP has been found to antagonize IL-22 binding and signaling. IL-22 appears not to directly influence immune cells, and major targets of the cytokine appear to be nonimmune cells, such as cells of the skin, digestive and respiratory system, as well as hepatocytes, and keratinocytes. IL-22 has been described as an effector cytokine of the Th17 lineage. Along with IL-17A and IL-17F, IL-22 regulates genes associated with innate immunity of the skin. IL-17A, IL-17F and IL-22 are all co-expressed by Th17 cells, however, they are differentially regulated. The effects of IL-22 include induction of acute phase reactants and antimicrobial proteins, as well as increasing the mobility of keratinocytes. IL-22 is highly expressed during chronic inflammation, and found to activate intracellular kinases and transcription factors. IL-22 is critical for host defense against infections of extracellular pathogens, and promotes wound-healing responses. IL-22 is upregulated in activated T cells. IL-22 has been reported to mediate IL-23-induced acanthosis and dermal inflammation through activation of STAT3.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso Q9GZX6
Para utilizar con (aplicación) ELISA, Functional Assay, Western Blot
Formulación protein with no preservative
ID de gen (Entrez) 50616
Peso molecular 33.6 kDa
Nombre Human IL-22
Cantidad 10 μg
Fuente E. coli
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen Cytokine Zcyto18; EGK_03903; il 22; IL-10-related T-cell-derived inducible factor; IL-10-related T-cell-derived-inducible factor; IL-21; IL22; IL-22; Il22a; IL-22 A; IL-D110; ILN; ILTIF; IL-TIF; ILTIF alpha; IL-TIF alpha; ILTIFa; Interleukin; interleukin 10-related T cell-derived inducible factor; interleukin 22; Interleukin22; interleukin-22; Interleukin-22 A; RGD1561292; TIFa; TIFIL-23; UNQ3099/PRO10096; ZCYTO18
Nombre común IL-22
Símbolo de gen IL22
Actividad biológica Assay 1: Determined by its ability to activate STAT following receptor ligand interaction. Assay 2: Measured by its ability to induce IL-10 secretion in COLO 205 (human colon carcinoma cells). The expected ED50 for this effect is 0.7- 1.0 ng/ml
Conjugado Unconjugated
Recombinante Recombinant
Secuencia MAPISSHCRL DKSNFQQPYI TNRTFMLAKE ASLADNNTDV RLIGEKLFHG VSMSERCYLM KQVLNFTLEE VLFPQSDRFQ PYMQEVVPFL ARLSNRLSTC HIEGDDLHIQ RNVQKLKDTV KKLGESGEIK AIGELDLLFM SLRNACI
Contenido y almacenamiento -20°C
Sistema de expresión E. coli
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.