missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IGF-I Recombinant Protein

Código de producto. 11584940
Change view
Click to view available options
Cantidad:
1000 μg
Tamaño de la unidad:
1000 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11584940 1000 μg 1000 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11584940 Proveedor Invitrogen™ N.º de proveedor undefined

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

This is a recombinant protein suitable for use as a control in various applications. Lyophilized product is stable at room temperature for 3 weeks. Store lyophilized product desiccated at -20°C. Upon reconstitution, store at 4°C for 2-7 days or aliquot and store at -20°C. For long-term storage add either HSA or BSA to 0.1%. Avoid repeated freeze-thaw cycles. Reconstitute in sterile water not less than 100 μg/mL.

IGF1 (Insulin-like growth factor-1) is structurally and functionally related to insulin but has a much higher growth-promoting activity. A variety of cellular responses are induced by IGF1, including cell proliferation, differentiation, migration, and survival. Further, IGF1 is a polypeptide growth factor that stimulates the proliferation of a wide range of cell types in muscle, bone, and cartilage tissue. IGF1 stimulates glucose transport in rat bone-derived osteoblastic (PyMS) cells and is effective at much lower concentrations than insulin, not only regarding glycogen and DNA synthesis but also with regards to enhancing glucose uptake. In circulation, IGFs are predominantly bound to binding proteins (IGFBPs) which prolong the half-life of the IGFs and play a role in delivering them to target tissues. IGF-I is known as one of the most potent activators of the AKT signaling pathway which is known to be a stimulator of proliferation and an inhibitor of programmed cell death. Moreover, mature human IGF-I is 100% homologous with bovine and porcine proteins. Low levels of IGF1 have been linked to Alzheimer's disease. IGF1 is processed from a precursor, bound by a specific receptor, and secreted. Defects in the IGF1 gene are a cause of insulin-like growth factor 1 deficiency and several transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P05019
Para utilizar con (aplicación) Bioactivity
Formulación PBS with no preservative
ID de gen (Entrez) 3479
Peso molecular 7.6 kDa
Nombre Human IGF-I
Método de purificación Chromatography
Cantidad 1000 μg
Fuente E. coli
Inmunógeno N/A
Requisitos de almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Alias de gen C730016P09Rik; class 1 insulin-like growth factor I preproprotein; class 2 insulin-like growth factor I preproprotein; H-IGF-1; IBP1; IGF; IGF1; Igf-1; IGF1A; IGFI; Igf-I; IGFIa; IGF-IA; IGF-IB; insulin growth factor-1; Insulin like growth factor; insulin like growth factor 1; insulin-like growth factor 1; insulin-like growth factor 1 (somatomedin C); insulin-like growth factor I; insulin-like growth factor I (somatomedin C); insulin-like growth factor IB; mechano growth factor; MGF; M-IGF-1; OTTHUMP00000195084; prepro-IGF-I; prepro-insulin-like growth factor I; R-IGF-1; somatomedin; Somatomedin C; Somatomedin-C
Nombre común IGF1
Símbolo de gen IGF1
Actividad biológica ED50 < 1.0 ng/mL; determined by the dose-dependent proliferation of mouse Balb/c 3T3 cells.
Tipo de producto Protein
Conjugado Unconjugated
Especie E. coli
Recombinante Recombinant
Secuencia Human IGF-1 recombinant protein contains 70 amino acids
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Actividad ED50 < 1.0 ng/mL; determined by the dose-dependent proliferation of mouse Balb/c 3T3 cells.
Sistema de expresión E. coli
Formulario Lyophilized
Subtipo de proteína Recombinant
Grado de pureza o calidad >97% by SDS-PAGE and analyzed by silver stain.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.