missing translation for 'onlineSavingsMsg'
Learn More

Human IgE Antibody (F4-#16), Novus Biologicals™

Product Code. p-200070911 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.025 mg
Unit Size:
0.03 miligramo
Product Code. Cantidad unitSize
18732994 0.025 mg 0.03 miligramo
1 options
This item is not returnable. View return policy

Product Code. 18732994

Brand: Novus Biologicals NBP1785780.025mg

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Human IgE Monoclonal specifically detects Human IgE in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Radioimmunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Human IgE
Aplicaciones Western Blot, ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
Clasificación Monoclonal
Clon F4-#16
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:500 - 1:2000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000
Alias de gen immunoglobulin heavy constant epsilon
Especie del huésped Mouse
Inmunógeno Human Myeloma IgE
Método de purificación Affinity Purified
Cantidad 0.025 mg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.