missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IER2 (aa 132-189) Control Fragment Recombinant Protein
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66339 (PA5-66339. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
DNA-binding protein that seems to act as a transcription factor (PubMed:19584537). Involved in the regulation of neuronal differentiation, acts upon JNK-signaling pathway activation and plays a role in neurite outgrowth in hippocampal cells. May mediate with FIBP FGF-signaling in the establishment of laterality in the embryo. Promotes cell motility, seems to stimulate tumor metastasis (PubMed:22120713). [UniProt]
Especificaciones
Especificaciones
| Número de acceso | Q9BTL4 |
| Concentración | ≥5.0 mg/mL |
| Para utilizar con (aplicación) | Blocking Assay, Control |
| Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
| ID de gen (Entrez) | 9592 |
| Nombre | Human IER2 (aa 132-189) Control Fragment |
| Cantidad | 100 μL |
| Estado normativo | RUO |
| Alias de gen | AI317238; Ch1; CH x 1; chxI protein homolog; cycloheximide-induced gene protein; ETR101; Growth factor-inducible immediate early protein; Ier2; immediate early response 2; immediate early response gene 2 protein; PIP92; proline-rich-induced protein 92; Protein ETR101; T-lymphocyte-activated protein |
| Nombre común | IER2 |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido