missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GRINA (aa 31-74) Control Fragment Recombinant Protein

Código de producto. 30204898
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30204898 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30204898 Proveedor Invitrogen™ N.º de proveedor RP96582

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57896 (PA5-57896. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The transmembrane BAX inhibitor motif (TMBIM) family of proteins includes the founder member TMBIM6/BI-1, TMBIM1/RECS1 (responsive to centrifugal force and shear stress gene 1 protein), TMBIM2/LFG (life guard), TMBIM3/GRINA (glutamate receptor ionotropic NMDA protein 1), TMBIM4/GAAP (Golgi anti-apoptotic-associated protein), and TMBIM5/GHTIM (growth hormone-inducible transmembrane protein). They are highly conserved in mammals and zebrafish and contain a conserved BAX inhibitor-1 motif. GRINA is expressed in the brain and is a potential apoptotic regulator.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso Q7Z429
Concentración ≥5.0 mg/mL
Para utilizar con (aplicación) Blocking Assay, Control
Formulación 1 M urea, PBS with no preservative; pH 7.4
ID de gen (Entrez) 2907
Nombre Human GRINA (aa 31-74) Control Fragment
Cantidad 100 μL
Estado normativo RUO
Alias de gen 1110025J15Rik; Glutamate [NMDA] receptor-associated protein 1; glutamate ionotropic receptor NMDA type subunit associated protein 1; glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1 (glutamate binding); glutamate receptor, NMDA subtype, glutamate-binding subunit; GRINA; HNRGW; Lag; LFG1; NMDA receptor glutamate-binding chain; NMDA receptor glutamate-binding subunit; Nmdara1; N-methyl-D-aspartate receptor glutamate-binding chain; Protein lifeguard 1; putative MAPK-activating protein PM02; TMBIM3; transmembrane BAX inhibitor motif containing 3; transmembrane BAX inhibitor motif-containing protein 3
Nombre común GRINA
Símbolo de gen GRINA
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína His-ABP-tag
Secuencia PYAQPPYPGAPYPQPPFQPSPYGQPGYPHGPSPYPQGGYPQGPY
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Sistema de expresión E. coli
Formulario Liquid
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.