missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human GDF-3, Animal-Free Recombinant Protein, PeproTech®
Click to view available options
Cantidad:
250 μg
50 μg
5 μg
Tamaño de la unidad:
250 microgramos
5 microgramos
50 microgramos
Descripción
Recombinant Human GDF-3 is a 26.0 kDa non-disulfide-linked homodimer containing two 114 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
GDF3 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. The function of this protein is unknown, but expression studies suggest it may be involved in regulation of the adult lymphatic and erythroid systems and embryonic development.
Especificaciones
Especificaciones
| Número de acceso | Q9NR23 |
| Para utilizar con (aplicación) | Functional Assay |
| Formato | Lyophilized |
| Formulación | protein with no preservative |
| ID de gen (Entrez) | 9573 |
| Peso molecular | 26 kDa |
| Nombre | Human GDF-3, Animal-Free |
| Método de purificación | HPLC, SDS-PAGE |
| Cantidad | 250 μg |
| Estado normativo | RUO |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido