Learn More
Invitrogen™ Human GAS6 (aa 503-633) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (76%), Rat (76%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52539. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Número de acceso | Q14393 |
| Concentración | 8.50 mg/mL |
| Para utilizar con (aplicación) | Neutralization, Control |
| Formulación | PBS, 1M urea with no preservative; pH 7.4 |
| ID de gen (Entrez) | 2621 |
| Nombre | Human GAS6 (aa 503-633) Control Fragment |
| Intervalo de pH | 7.4 |
| Método de purificación | Purified |
| Cantidad | 100 μL |
| Requisitos de almacenamiento | -20°C, Avoid Freeze/Thaw Cycles |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.