missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Fractalkine (CX3CL1) Recombinant Protein, PeproTech®

Código de producto. 17842553
Change view
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
250 μg
500 μg
50 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
5 microgramos
50 microgramos
500 microgramos
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17842553 100 μg 100 microgramos
17872283 1 mg 1 miligramo
17862953 20 μg 20 microgramos
17882483 250 μg 250 microgramos
17822593 500 μg 500 microgramos
17802163 50 μg 50 microgramos
17802893 5 μg 5 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17842553 Proveedor Gibco™ N.º de proveedor 30031100UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Human Fractalkine is an 8.5 kDa protein containing 76 amino acid residues, including the four conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Chemokines are a family of proteins associated with the trafficking of leukocytes in immune surveillance and inflammatory cell recruitment. They are classified based on the positions of key cysteine residues. CX3CL1 is a CX3C chemokine known to induce adhesion and migration of leukocytes mediated by a membrane-bound and soluble form respectively. Recent experiments have shown that CX3CL1 can suppress the production of nitrous oxide, interleukin-6, and TNF-a in activated microglia and neuronal cells, suggesting that it may act as an intrinsic inhibitor against neurotoxicity by activated microglia. Its receptor, CX3CR1, also functions as a co-receptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by CX3CL1.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P78423
Para utilizar con (aplicación) ELISA, Functional Assay, Western Blot
Formulación protein with no preservative
ID de gen (Entrez) 6376
Peso molecular 8.5 kDa
Nombre Human Fractalkine (CX3CL1)
Cantidad 100 μg
Fuente E. coli
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen A-152E5.2; AB030188; ABCD-3; Acc1; AI848747; C3Xkine; chemokine (C-X3-C motif) ligand 1; CX3C; CX3C membrane-anchored chemokine; C-X3-C motif chemokine 1; C-X3-C motif chemokine ligand 1; Cx3cl1; CXC3; CXC3C; D8Bwg0439e; Fkn; Fractalkine; neurotactin; NTN; NTT; Processed fractalkine; SCYD1; small inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, neurotactin); small inducible cytokine subfamily D (Cys-X3-Cys), member-1; small inducible cytokine subfamily D, 1; small-inducible cytokine D1
Nombre común CX3CL1
Símbolo de gen Cx3cl1
Actividad biológica Determined by its ability to chemoattract human T cells using a concentration range of 5.0-10.0 ng/ml
Conjugado Unconjugated
Recombinante Recombinant
Secuencia QHHGVTKCNI TCSKMTSKIP VALLIHYQQN QASCGKRAII LETRQHRLFC ADPKEQWVKD AMQHLDRQAA ALTRNG
Contenido y almacenamiento -20°C
Sistema de expresión E. coli
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.