missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Flt-3 Ligand (FLT3L) Recombinant Protein, PeproTech®
Description
Recombinant Human Flt3-Ligand is a soluble 17.6 kDa protein consisting of 155 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
FLT3LG (Fms Related Tyrosine Kinase 3 Ligand) is a Protein Coding gene. Diseases associated with FLT3LG include Aplastic Anemia and Brain Cancer. Among its related pathways are Hematopoietic Stem Cell Differentiation Pathways and Lineage-specific Markers and Ras signaling pathway. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and cytokine activity. Dendritic cells (DCs) provide the key link between innate and adaptive immunity by recognizing pathogens and priming pathogen-specific immune responses. FLT3LG controls the development of DCs and is particularly important for plasmacytoid DCs and CD8-positive classical DCs and their CD103-positive tissue counterparts.
Spécification
Spécification
| Número de acceso | P49771 |
| Para utilizar con (aplicación) | ELISA, Functional Assay, Western Blot |
| Formulación | protein with no preservative |
| ID de gen (Entrez) | 2323 |
| Peso molecular | 17.6 kDa |
| Nombre | Human Flt-3 Ligand (FLT3L) |
| Cantidad | 250 μg |
| Fuente | E. coli |
| Estado normativo | RUO |
| Concentración de endotoxinas | <1 EU/ μg |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?