missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF basic/FGF2/bFGF Antibody, Novus Biologicals™

Rat Monoclonal Antibody
Marca: R&D Systems MAB2332-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
FGF basic/FGF2/bFGF Monoclonal antibody specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry
Especificaciones
| FGF basic/FGF2/bFGF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin | |
| E. coli-derived human FGF basic/FGF2/bFGF, Pro143-Ser288, Accession # P09038 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry, Immunocytochemistry | |
| 954824 | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P09038 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 2247 | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido