missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FGF-10, Recombinant Protein
Descripción
FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This protein is also implicated to be a primary factor in the process of wound healing.
Especificaciones
Especificaciones
| Número de acceso | O15520 |
| Para utilizar con (aplicación) | Control, ELISA, Immunohistochemistry, Western Blot |
| Formulación | protein with 0.02% Tween 80, 140 mM NaCl, 2.7 mM KCl, 5% mannitol, 5% trehalose and no preservative; pH 7.4 |
| ID de gen (Entrez) | 2255 |
| Nombre | Human FGF-10 |
| Cantidad | 5 μg |
| Fuente | E. coli |
| Requisitos de almacenamiento | -20°C |
| Estado normativo | RUO |
| Alias de gen | AEY17; BB213776; FGF; FGF10; FGF-10; Fibroblast growth factor; Fibroblast growth factor 10; Gsfaey17; Keratinocyte growth factor 2; produced by fibroblasts of urinary bladder lamina propria |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?