missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Fetuin A/AHSG Recombinant Protein, PeproTech®

Código de producto. 17850563
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
250 μg
500 μg
50 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
250 microgramos
50 microgramos
500 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17850563

Marca: Gibco™ 14013500UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Human Fetuin A is a heterodimeric protein containing a 282 amino acid A chain and a 27 amino acid B chain. The calculated molecular weight of Recombinant Human Fetuin A is 32.9 kDa. Due to glycosylation, recombinant Fetuin A migrates at an apparent molecular weight of 45-55 kDa by SDS-PAGE run under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Alpha2-HS glycoprotein (AHSG), a glycoprotein present in the serum, is synthesized by hepatocytes. The AHSG molecule consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues. However, its exact significance is still obscure.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P02765
Para utilizar con (aplicación) Functional Assay
Formulación protein with no preservative
ID de gen (Entrez) 197
Peso molecular 32.9 kDa
Nombre Human Fetuin A/AHSG
Cantidad 500 μg
Fuente HEK293 cells
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen 59 kDa bone sialic acid-containing protein; A2HS; Aa2-066; AHS; AHSG; alpha 2 HS-glycoprotein alpha 2 (fetuin); alpha 2-HS glycoprotein; alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; asialofetuin; Ba-alpha-2-glycoprotein; BSP; Countertrypin; FETUA; fetuin; fetuin A; FetuinA; fetuin-A; Glycoprotein PP63; HSGA; phosphorylated N-glycoprotein pp63; pp63; PRO2743
Nombre común Fetuin A
Símbolo de gen AHSG
Actividad biológica Determined by its ability to inhibit Cathespin V cleavage of a fluorogenic peptide substrate Z-LR-AMC (RLUs). The expected IC50 is ≤ 100 nM.
Conjugado Unconjugated
Recombinante Recombinant
Secuencia A-Chain: APHGPGLIYR QPNCDDPETE EAALVAIDYI NQNLPWGYKH TLNQIDEVKV WPQQPSGELF EIEIDTLETT CHVLDPTPVA RCSVRQLKEH AVEGDCDFQL LKLDGKFSVV YAKCDSSPDS AEDVRKVCQD CPLLAPLNDT RVVHAAKAAL AAFNAQNNGS NFQLEEISRA QLVPLPPSTY VEFTVSGTDC VAKEATEAAK CNLLAEKQYG FCKATLSEKL GGAEVAVTCT VFQTQPVTSQ PQPEGANEAV PTPVVDPDAP PSPPLGAPGL PPAGSPPDSH VL B-Chain: TVVQPSVG AAAGPVVPPC PGRIRHFKV
Contenido y almacenamiento -20°C
Sistema de expresión HEK293 cells
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado