missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FAM63B (aa 80-145) Control Fragment Recombinant Protein

Código de producto. 30194292
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
30194292 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. View return policy

Product Code. 30194292

Brand: Invitrogen™ RP107273

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (50%), Rat (50%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66614 (PA5-66614. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Hydrolase that can remove 'Lys-48'-linked conjugated ubiquitin from proteins (PubMed:27292798). Binds to polyubiquitin chains of different linkage types, including 'Lys-6', 'Lys-11', 'Lys-29', 'Lys-33', 'Lys-48' and 'Lys-63' (PubMed:28082312). May play a regulatory role at the level of protein turnover (PubMed:27292798). [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Número de acceso Q8NBR6
Concentración ≥5.0 mg/mL
Para utilizar con (aplicación) Blocking Assay, Control
Formulación 1 M urea, PBS with no preservative; pH 7.4
ID de gen (Entrez) 54629
Nombre Human FAM63B (aa 80-145) Control Fragment
Cantidad 100 μL
Estado normativo RUO
Alias de gen Deubiquitinating enzyme MINDY-2; FAM63B; family with sequence similarity 63 member B; family with sequence similarity 63, member B; KIAA1164; MINDY2; Protein FAM63B; Ubiquitin carboxyl-terminal hydrolase MINDY-2
Nombre común FAM63B
Símbolo de gen MINDY2
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína His-ABP-tag
Secuencia SSSAGLDLKDSGLESPAAAEAPLRGQYKVTASPETAVAGVGHELGTAGDAGARPDLAGTCQAELTA
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Sistema de expresión E. coli
Formulario Liquid
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.