missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Epiregulin, Animal-Free Recombinant Protein, PeproTech®
Descripción
Recombinant Human Epiregulin is a 5.6 kDa monomeric protein, containing 50 amino residues, which corresponds to the mature secreted Epiregulin sequence. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Epiregulin is an EGF-related growth factor that binds specifically to EGFR (ErbB1) and ErbB4, but not ErbB2 or ErbB3. It is expressed mainly in the placenta and peripheral blood leukocytes, as well as in certain carcinomas of the bladder, lung, kidney and colon. Epiregulin stimulates the proliferation of keratinocytes, hepatocytes, fibroblasts and vascular smooth muscle cells. It also inhibits the growth of several tumor-derived epithelial cell lines. Human Epiregulin is initially synthesized as a glycosylated 19.0 kDa transmembrane precursor protein, which is processed by proteolytic cleavage to produce a 6.0 kDa mature secreted sequence.
Especificaciones
Especificaciones
| Número de acceso | O14944 |
| Para utilizar con (aplicación) | Functional Assay |
| Formato | Lyophilized |
| Formulación | protein with no preservative |
| ID de gen (Entrez) | 2069 |
| Peso molecular | 5.6 kDa |
| Nombre | Human Epiregulin, Animal-Free |
| Método de purificación | HPLC, SDS-PAGE |
| Cantidad | 5 μg |
| Estado normativo | RUO |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?