missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CXCL9 (MIG) Recombinant Protein, PeproTech®

Código de producto. 17882353
Change view
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
250 μg
500 μg
50 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
5 microgramos
50 microgramos
500 microgramos
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17882353 100 μg 100 microgramos
17872353 1 mg 1 miligramo
17862253 20 μg 20 microgramos
17862763 250 μg 250 microgramos
17802853 500 μg 500 microgramos
17812493 50 μg 50 microgramos
17892343 5 μg 5 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17882353 Proveedor Gibco™ N.º de proveedor 30026100UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Human MIG is an 11.7 kDa protein containing 103 amino acid residues, including the four highly conserved cysteine residues present in CXC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

MIG, also known as CXCL9, is a T-cell chemoattractant inducible by gamma interferon that is a member of the CXC chemokine family of cytokines.This gene is mapped to 4q21. It is noted that, although the best-described activities of the chemokines are as chemotactic factors, chemokines also have an effect on T-cell activation, angiogenesis, and HIV infection. While most CXC chemokines are chemotactic for neutrophils, MIG and INP10 are unusual and similar in being CXC chemokines that are chemotactic for lymphocytes and inactive in neutrophils.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso Q07325
Para utilizar con (aplicación) ELISA, Functional Assay, Western Blot
Formulación protein with no preservative
ID de gen (Entrez) 4283
Peso molecular 11.7 kDa
Nombre Human CXCL9 (MIG)
Cantidad 100 μg
Fuente E. coli
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen BB139920; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 9; CMK; crg-10; CXC; CXC motif chemokine; C-X-C motif chemokine 9; C-X-C motif chemokine ligand 9; CXCL; CXCL9; gamma interferon-induced monokine; gamma-interferon-induced monokine; HuMIG; M119; Mig; monokine induced by gamma interferon; monokine induced by interferon gamma; monokine induced by interferon-gamma; MuMIG; protein m119; SCYB9; small inducible cytokine B subfamily (Cys-X-Cys), member 9; small inducible cytokine B9; small-inducible cytokine B9
Nombre común CXCL9 (MIG)
Símbolo de gen CXCL9
Actividad biológica Determined by its ability to chemoattract human peripheral blood T lymphocytes using a concentration range of 10.0-100.0 ng/ml
Conjugado Unconjugated
Recombinante Recombinant
Secuencia TPVVRKGRCS CISTNQGTIH LQSLKDLKQF APSPSCEKIE IIATLKNGVQ TCLNPDSADV KELIKKWEKQ VSQKKKQKNG KKHQKKKVLK VRKSQRSRQK KTT
Contenido y almacenamiento -20°C
Sistema de expresión E. coli
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.