missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CXCL12 (SDF-1g) Recombinant Protein, PeproTech®

Código de producto. 17889375
Change view
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
250 μg
2 μg
500 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
2 microgramos
250 microgramos
500 microgramos
Código de producto. Cantidad unitSize
17889375 2 μg 2 microgramos
17829285 500 μg 500 microgramos
17869375 1 mg 1 miligramo
17899245 10 μg 10 microgramos
17889665 100 μg 100 microgramos
17809555 250 μg 250 microgramos
6 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17889375

Proveedor: Gibco™ 30028G2UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Human CXCL12 (SDF-1g) is a 11.6 kDa protein containing 98 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The SDF-1 proteins are stromal-derived CXC chemokines that signal primarily through the CXCR4 receptor. SDF-1 proteins exert chemotactic activity over a broad range of immune system cell types and may also be involved in other processes including metastasis of certain cancers, inflammation, and angiogenesis. Additionally, SDF-1 can suppress HIV entry into cells expressing the CXCR4 receptor. Five isomers of SDF-1 have been found with SDF-1alpha being the predominant form, expressing in a wide range of cell types. The SDF-1gamma isoform is distinctive in that it contains a highly basic C-terminal region, which presumably is the reason for its high binding affinity to gylcosaminoglycans (GAG) and heparin sulfate (HS). Consequently, most of SDF-1gamma exists in a cell- and membrane-bound form, which confers greater stability compared to other isoforms via increased resistance to proteolytic degradation. SDF-1gamma binds with reduced affinity to the CXCR4 receptor when compared to the alpha and beta isoforms, but because of its increased stability can, over time, exert equal or greater activity when compared to other SDF-1 isoforms. SDF-1gamma expression has been found primarily in the heart (human and mouse) and in the brain and central nervous system (rat).
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P48061
Para utilizar con (aplicación) Functional Assay
Formato Lyophilized
Formulación protein with no preservative
ID de gen (Entrez) 6387
Peso molecular 11.6 kDa
Nombre Human CXCL12 (SDF-1 g)
Método de purificación HPLC, SDS-PAGE
Cantidad 2 μg
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen PBSF; Stromal-Cell Derived Factor-1
Nombre común CXCL12 gamma
Símbolo de gen CXCL12
Actividad biológica Determined by its ability to chemoattract human T cells.
Tipo de producto Protein
Conjugado Unconjugated
Recombinante Recombinant
Secuencia KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKGR REEKVGKKEK IGKKKRQKKR KAAQKRKN
Contenido y almacenamiento -20°C
Actividad Determined by its ability to chemoattract human T cells.
Sistema de expresión E. coli
Formulario Lyophilized
Grado de pureza o calidad ≥98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.