missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CWC22 Control Fragment Recombinant Protein
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (46%), Rat (46%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57796 (PA5-57796. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Required for pre-mRNA splicing as component of the spliceosome (PubMed:12226669, PubMed:11991638, PubMed:22961380, PubMed:28502770, PubMed:28076346, PubMed:29360106, PubMed:29301961). Promotes exon-junction complex (EJC) assembly (PubMed:22959432, PubMed:22961380). Hinders EIF4A3 from non-specifically binding RNA and escorts it to the splicing machinery to promote EJC assembly on mature mRNAs. Through its role in EJC assembly, required for nonsense-mediated mRNA decay. [UniProt]
Especificaciones
Especificaciones
| Número de acceso | Q9HCG8 |
| Concentración | ≥5.0 mg/mL |
| Para utilizar con (aplicación) | Blocking Assay, Control |
| Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
| ID de gen (Entrez) | 57703 |
| Nombre | Human CWC22 Control Fragment |
| Cantidad | 100 μL |
| Estado normativo | RUO |
| Alias de gen | AA684037; AI173004; AL022752; B230213M24; Cwc22; CWC22 homolog, spliceosome-associated protein; CWC22 spliceosome-associated protein; CWC22 spliceosome-associated protein homolog; CWC22 spliceosome-associated protein homolog (S. cerevisiae); EIF4GL; fSAPb; functional spliceosome-associated protein b; KIAA1604; mKIAA1604; NCM; Nucampholin homolog; pre-mRNA-splicing factor CWC22 homolog; RGD1565385 |
| Nombre común | CWC22 |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido