missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coronavirus Spike Protein Antibody (028)
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP2-90334-100ul
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Human Coronavirus Spike Protein Monoclonal specifically detects Human Coronavirus Spike Protein in Virus samples. It is validated for ELISA.
Especificaciones
| Human Coronavirus Spike Protein | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| HKU1, Spike glycoprotein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant HCoV-HKU1 (isolate N1) Spike/S1 (YP_173238.1; Met1-Arg760) | |
| 100 μL | |
| Primary | |
| HCoV-HKU1 (isolate N1) Spike/S1 No cross-reactivity in ELISA with: HCoV-HKU1 (isolate N5) Spike S1+S2 ECD Protein SARS-CoV-2 Spike S1 Protein SARS-CoV-2 Spike S1 NTD Protein SARS-CoV-2 Spike S1 NTD Protein SARS-CoV-2 Spike RBD Protein SARS-CoV-2 Spike S2 Protein SARS-CoV-2 Spike S1+S2 ECD Protein SARS-CoV Spike S1+S2 ECD (S577A, Isolate Tor2) Protein MERS-CoV Spike S1+S2 ECD Protein HCoV-NL63 Spike S1+S2 ECD Protein HCoV-229E Spike S1+S2 ECD Protein HCoV-OC43 Spike S1+S2 ECD Protein | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 28 | |
| ELISA 1:5000-1:10000 | |
| YP_173238.1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3200426 | |
| Virus | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido