missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CDKN2B (aa 89-122) Control Fragment Recombinant Protein
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-145041 (PA5-145041. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The four known 15-to 19 kDa INK4 proteins (p15INK4b, p16INK4a, p18INK4c andp19INK4d) bind and inhibit CDK4 and CDK6 but not other CDKs, including cyclin E-Cdk2, cyclin A-Cdk2 and cyclin B-Cdk1. Like the three D-type cyclins, the INK4 genes are expressed in distinct tissue-specific patterns, suggesting that they are not strictly redundant. 5 p18INK4c is expressed during G1 to S transition in the eukaryotic cell division cycle. It is maximally induced as cells enter S-phase.
Especificaciones
Especificaciones
| Número de acceso | P42772 |
| Concentración | ≥5.0 mg/mL |
| Para utilizar con (aplicación) | Blocking Assay, Control |
| Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
| ID de gen (Entrez) | 1030 |
| Nombre | Human CDKN2B (aa 89-122) Control Fragment |
| Cantidad | 100 μL |
| Estado normativo | RUO |
| Alias de gen | AV083695; CDK inhibitory protein; CDK4B inhibitor; CDK4I; Cdkn2b; cyclin dependent kinase inhibitor 2 B; Cyclin dependent kinase inhibitor 2 B (p15, inhibits CDK4); cyclin-dependent kinase 4 inhibitor B; cyclin-dependent kinase inhibitor 2 B (p15, inhibits CDK4); cyclin-dependent kinase inhibitor p15; cyclin-dependent kinase inhibitor p15INK4b; cyclin-dependent kinase inhibitor protein; cyclin-dependent kinases 4 and 6 binding protein; Ink4; INK4B; MTS2; MTS-2; Multiple tumor suppressor 2; p14_CDK inhibitor; p14_INK4B; p14-INK4b; P15; p15 CDK inhibitor; p15(INK4b); p15_INK4B; p15INK4b; p15-INK4b; TP15 |
| Nombre común | CDKN2B |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido