missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CDK5RAP3 (aa 170-262) Control Fragment Recombinant Protein
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54627 (PA5-54627. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a protein that has been reported to function in signaling pathways governing transcriptional regulation and cell cycle progression. It may play a role in tumorigenesis and metastasis. A pseudogene of this gene is located on the long arm of chromosome 20. Alternative splicing results in multiple transcript variants that encode different isoforms. [provided by RefSeq, May 2013].
Especificaciones
Especificaciones
| Número de acceso | Q96JB5 |
| Concentración | ≥5.0 mg/mL |
| Para utilizar con (aplicación) | Blocking Assay, Control |
| Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
| ID de gen (Entrez) | 80279 |
| Nombre | Human CDK5RAP3 (aa 170-262) Control Fragment |
| Cantidad | 100 μL |
| Estado normativo | RUO |
| Alias de gen | 1810007E24Rik; BC002318; C53; C81486; CDK5 activator-binding protein C53; CDK5 activator-binding protein C53; CDK5 regulatory subunit associated protein 3; CDK5 regulatory subunit associated protein IC53-2; CDK5 regulatory subunit-associated protein 3; Cdk5rap3; HSF-27; IC53; IC53-2; ischemic heart CDK5 activator-binding protein C53; LXXLL/leucine-zipper-containing ARFbinding protein; LXXLL/leucine-zipper-containing ARF-binding protein; LZAP; MST016; MSTP016; OK/SW-cl0.114; PP1553; Protein HSF-27 |
| Nombre común | CDK5RAP3 |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido