Learn More
Invitrogen™ Human CD3 epsilon Heterodimer Recombinant Protein
Código de producto. p-200008576
Click to view available options
Cantidad:
500 μg
50 μg
Tamaño de la unidad:
50 microgramos
500 microgramos
Código de producto. 17906411
Marca: Invitrogen™ RP87999
Recombinant Protein
It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (200 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.
CD3E and CD3D Heterodimer Protein is produced by mammalian expression system and are involved in mediating signal transduction.
TRUSTED_SUSTAINABILITY
Especificaciones
Número de acceso P04234, P07766
Concentración 0.25 mg/mL
Para utilizar con (aplicación) Control, ELISA Standard, Binding Assay
Formato Lyophilized
Formulación PBS with 5% mannitol, 5% trehalose and no preservative; pH 7.4
ID de gen (Entrez) 915, 916
Peso molecular 77.5 kDa
Nombre Human CD3 epsilon Heterodimer
Intervalo de pH 7.4
Método de purificación Purified
Cantidad 500 μg
Requisitos de almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Concentración de endotoxinas <1.0 EU/μg
Nombre común CD3e/CD3d
Símbolo de gen CD3D, CD3E
Actividad biológica Measured by its ability to bind OKT3-mIgG2a in a functional ELISA.
Tipo de producto Protein
Conjugado Unconjugated
Recombinante Recombinant
Etiqueta de proteína Fc-tag
Secuencia Human CD3D, Met1-Ala105 (Accession # P04234-1) with a C-terminal His-tag hIgG1 Fc region and human CD3E, Met1-Asp126 (Accession # P07766-1) with a C-terminal Flag-tag hIgG Fc region were co-expressed and purified as a human CD3D & CD3E heterodimer
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Actividad Measured by its ability to bind OKT3-mIgG2a in a functional ELISA.
Sistema de expresión HEK293 cells
Formulario Lyophilized
Grado de pureza o calidad ≥90% as determined by SDS-PAGE. ≥95% as determined by SEC-HPLC.