missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CCL11 (Eotaxin) Recombinant Protein, PeproTech®

Código de producto. 17872853
Change view
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
250 μg
500 μg
50 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
5 microgramos
50 microgramos
500 microgramos
Código de producto. Cantidad unitSize
17872853 250 μg 250 microgramos
17842463 100 μg 100 microgramos
17892383 1 mg 1 miligramo
17822193 20 μg 20 microgramos
17892073 500 μg 500 microgramos
17872053 50 μg 50 microgramos
17822083 5 μg 5 microgramos
7 options
This item is not returnable. View return policy

Product Code. 17872853

Brand: Gibco™ 30021250UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Eotaxin is an 8.3 kDa protein containing 74 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
TRUSTED_SUSTAINABILITY

Specifications

Número de acceso P51671
Para utilizar con (aplicación) ELISA, Functional Assay, Western Blot
Formulación protein with no preservative
ID de gen (Entrez) 6356
Peso molecular 8.3 kDa
Nombre Human CCL11 (Eotaxin)
Cantidad 250 μg
Fuente E. coli
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen C-C motif chemokine 11; C-C motif chemokine ligand 11; Ccl11; chemokine (C-C motif) ligand 11; eosinophil chemotactic protein; Eotaxin; Eotaxin 1; Eotaxin1; eotaxin-1; MGC22554; Scya11; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin); small inducible cytokine subfamily A11; small-inducible cytokine A11
Nombre común Eotaxin
Símbolo de gen CCL11
Actividad biológica Determined by its ability to chemoattract human peripheral blood eosinophils using a concentration range of 0.1-10.0 ng/ml
Conjugado Unconjugated
Recombinante Recombinant
Secuencia GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP
Contenido y almacenamiento -20°C
Sistema de expresión E. coli
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.