missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CCDC169 (aa 78-137) Control Fragment Recombinant Protein
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (33%), Rat (33%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144790 (PA5-144790. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Comprising nearly 4% of human DNA, chromosome 13 contains around 114 million base pairs and 400 genes. Key tumor suppressor genes on chromosome 13 include the breast cancer susceptibility gene, BRCA2, and the RB1 (retinoblastoma) gene. RB1 encodes a crucial tumor suppressor protein which, when defective, leads to malignant growth in the retina and has been implicated in a variety of other cancers. The gene SLITRK1, which is associated with Tourette syndrome, is on chromosome 13. As with most chromosomes, polysomy of part or all of chromosome 13 is deleterious to development and decreases the odds of survival. Trisomy 13, also known as Patau syndrome, is quite deadly and the few who survive past one year suffer from permanent neurologic defects, difficulty eating and vulnerability to serious respiratory infections.
Especificaciones
Especificaciones
| Número de acceso | A6NNP5 |
| Concentración | ≥5.0 mg/mL |
| Para utilizar con (aplicación) | Blocking Assay, Control |
| Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
| ID de gen (Entrez) | 728591 |
| Nombre | Human CCDC169 (aa 78-137) Control Fragment |
| Cantidad | 100 μL |
| Estado normativo | RUO |
| Alias de gen | A730037C10Rik; C13orf38; CCDC169; coiled-coil domain containing 169; coiled-coil domain-containing protein 169; RP11-251J8.1 |
| Nombre común | CCDC169 |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido