missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ALK/CD246 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB4210-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ALK/CD246 Monoclonal specifically detects ALK/CD246 in Human samples. It is validated for Western Blot.
Especificaciones
| ALK/CD246 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9UM73 | |
| ALK | |
| Mouse myeloma cell line NS0-derived recombinant human ALK/CD246 Val19-Ser1038 Accession # Q9UM73 | |
| 25 μg | |
| Primary | |
| Detects human ALK/CD246 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 383625 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, Ki-1, NBLST3, Tcrz | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 238 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido