Learn More
HSPH1 Mouse anti-Human, Clone: 3H5A5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66723-1-IG-20UL
Descripción
Western blot analysis using this antibody detected a band around 110 kDa in various lysates.
HSPH1 prevents the aggregation of denatured proteins in cells under severe stress, on which the ATP levels decrease markedly. This protein inhibits HSPA8/HSC70 ATPase and chaperone activities.Especificaciones
| HSPH1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q92598 | |
| HSPH1 | |
| HSPH1 Fusion Protein Ag27521 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| 3H5A5 | |
| Unconjugated | |
| HSPH1 | |
| Antigen NY CO 25, DKFZp686M05240, Heat shock 110 kDa protein, Heat shock protein 105 kDa, HSP105, HSP105A, HSP105B, HSP110, HSPH1, KIAA0201, NY CO 25 | |
| Mouse | |
| Protein G | |
| RUO | |
| 10808 | |
| -20°C | |
| Liquid |