missing translation for 'onlineSavingsMsg'
Learn More

HSPC323 Antibody, Novus Biologicals™

Código de producto. p-200081712 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μL
25 μL
Tamaño de la unidad:
100 microlitros
25 microlitros
Código de producto. Cantidad unitSize
18250603 100 μL 100 microlitros
18619526 25 μL 25 microlitros
2 options
This item is not returnable. View return policy

Product Code. 18250603

Brand: Novus Biologicals NBP258735

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HSPC323 Polyclonal specifically detects HSPC323 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno HSPC323
Aplicaciones Immunocytochemistry, Immunofluorescence
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulación PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Alias de gen C19orf77, chromosome 19 open reading frame 77, HSPC323, transmembrane protein C19orf77, transmembrane protein HSPC323
Símbolos de los genes C19orf77
Especie del huésped Rabbit
Inmunógeno This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KTAKEGESNLGLDLEEKEPGDHERAKSTVM
Método de purificación Affinity Purified
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 284422
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.