missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
HRP Polyclonal antibody specifically detects HRP in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Especificaciones
Especificaciones
| Antígeno | HRP |
| Aplicaciones | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation |
| Clasificación | Polyclonal |
| Concentración | LYOPH |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:1000-1:4000, ELISA 1:10000-1:40000, Immunohistochemistry, Immunoprecipitation |
| Formulación | Lyophilized from 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 |
| Alias de gen | EC 1.11.1.7, Horseradish Peroxidase, HPRC 1, HPRC1, HRPA2, HRPC 2, HRPC2, HRPE5, HRPN, Neutral peroxidase, Peroxidase A2, Peroxidase C1A, Peroxidase C1B, Peroxidase C2, Peroxidase C3, Peroxidase E5, Peroxidase N, PRXC1A, PRXC1B, PRXC2, PRXC3 |
| Especie del huésped | Sheep |
| Inmunógeno | HRP [Horseradish] (Uniprot: Q42517) |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?