Learn More
HPSE Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24529-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Stem cells offer a therapeutic promise for many diseases. In the case of diabetes, stem cells offer the promise that beta cells may be generated ex vivo for the possible transplantation and cure of Type I diabetes (Couri CE, et al.). For this to happen, stem cells will need to be isolated and expanded, they will need to be differentiated toward the beta cell lineage, and the differentiated cell progeny used for transplantation may need to be isolated from complex cell mixtures following differentiation. Appropriate markers targeting cell surface molecules on developing and mature pancreatic cells will be required for such research.Especificaciones
| HPSE | |
| Polyclonal | |
| Unconjugated | |
| HPSE | |
| Endo glucoronidase, HEP, heparanase, Heparanase 1, HPA, Hpa1, HPR1, HPSE, HPSE1, HSE1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10855, 15442 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6YGZ1, Q9Y251 | |
| HPSE | |
| HPSE Fusion Protein Ag21479 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |